4-Bromheptan 97%

Lieferant: Apollo Scientific

OR2933-25G OR2933-5G
APOSOR2933-25GEA 323 EUR
APOSOR2933-25G APOSOR2933-5G
4-Bromheptan 97%
4-Bromheptan
Formel: H₃C(CH₂)₂BrCH(CH₂)₂CH₃
Molecular Weight: 179,1 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039182
CAS-Nummer: 998-93-6
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR