3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid

Lieferant: Apollo Scientific
Achtung

OR29368-1G OR29368-250MG
APOSOR29368-1GEA 185 EUR
APOSOR29368-1G APOSOR29368-250MG
3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid
3-(3,5-Dibromo-4-hydroxyphenyl)propanoic acid
Formel: C₉H₈Br₂O₃
Schmelzpunkt: 108…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00816760
CAS-Nummer: 13811-12-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR