2-(2-Chlorobenzylthio)ethylamine
Lieferant: Apollo Scientific
|
Gefahr
|
OR29429-1G
OR29429-250MG
APOSOR29429-1GEA
198
EUR
APOSOR29429-1G
APOSOR29429-250MG
2-(2-Chlorobenzylthio)ethylamine
2-(2-Chlorobenzylthio)ethylamine
Formel:
C₉H₁₂ClNS Molecular Weight: 201,712 g/mol Siedepunkt: 160…165 °C/13mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD01764494 CAS-Nummer: 42416-23-9 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...