2-(4-Pyridinyl)-4-thiazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

OR29448-1G OR29448-5G
APOSOR29448-1GEA 0 EUR
APOSOR29448-1G APOSOR29448-5G
2-(4-Pyridinyl)-4-thiazolcarbonsäure
2-(4-Pyridinyl)-4-thiazolcarbonsäure
Formel: C₉H₆N₂O₂S
Molecular Weight: 206.22 g/mol
Schmelzpunkt: >300 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00171745
CAS-Nummer: 21278-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR