3,5-Dichlorbenzylalkohol

Lieferant: Apollo Scientific
Achtung

OR29565-25G OR29565-5G OR29565-100G
APOSOR29565-25GEA 29 EUR
APOSOR29565-25G APOSOR29565-5G APOSOR29565-100G
3,5-Dichlorbenzylalkohol
3,5-Dichlorbenzylalkohol
Formel: C₇H₆Cl₂O
Molecular Weight: 177,03 g/mol
Schmelzpunkt: 76…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004634
CAS-Nummer: 60211-57-6
EINECS: 262-109-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR