(4-Methoxyphenoxy)acetonitril

Lieferant: Apollo Scientific

OR29620-10G OR29620-5G
APOSOR29620-10GEA 68.3 EUR
APOSOR29620-10G APOSOR29620-5G
(4-Methoxyphenoxy)acetonitril
(4-Methoxyphenoxy)acetonitril
Formel: C₉H₉NO₂
Molecular Weight: 163.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00017528
CAS-Nummer: 22446-12-4

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR