4-Methoxy-2-nitrobenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR2967-1G
OR2967-5G
APOSOR2967-1GEA
21.9
EUR
APOSOR2967-1G
APOSOR2967-5G
4-Methoxy-2-nitrobenzaldehyd 98%
4-Methoxy-2-nitrobenzaldehyde
Formel:
C₈H₇NO₄ Molecular Weight: 181,1455 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00024254 CAS-Nummer: 22996-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...