Methyl-3-methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylate

Lieferant: Apollo Scientific

OR29688-10G OR29688-1G OR29688-5G
APOSOR29688-10GEA 778 EUR
APOSOR29688-10G APOSOR29688-1G APOSOR29688-5G
Methyl-3-methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylate
Methyl-3-methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylate
Formel: C₁₀H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00085130
CAS-Nummer: 175277-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR