Methyl-4-(1-hydroxyiminoethyl)-5-methylisoxazole-3-carboxylate

Lieferant: Apollo Scientific

OR29703-1G OR29703-5G
APOSOR29703-1GEA 125 EUR
APOSOR29703-1G APOSOR29703-5G
Methyl-4-(1-hydroxyiminoethyl)-5-methylisoxazole-3-carboxylate
Methyl-4-(1-hydroxyiminoethyl)-5-methylisoxazole-3-carboxylate
Formel: C₈H₁₀N₂O₄
Molecular Weight: 198.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00068117
CAS-Nummer: 175277-20-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR