2,4-Dioxo-4-thiophen-2-ylbutyric acid ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,4-dioxo-4-(thiophen-2-yl)butanoate, Ethyl 2,4-dioxo-4-(thien-2-yl)butanoate

OR29711-1G OR29711-5G
APOSOR29711-1GEA 72.5 EUR
APOSOR29711-1G APOSOR29711-5G
2,4-Dioxo-4-thiophen-2-ylbutyric acid ethyl ester
2,4-Dioxo-4-thiophen-2-ylbutyric acid ethyl ester
Formel: C₁₀H₁₀O₄S
Molecular Weight: 226.25 g/mol
Schmelzpunkt: 34 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00126316
CAS-Nummer: 36983-36-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR