Ethyl-2,4-dichloro-6-methylpyridine-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2,4-dichloro-6-methyl-3-pyridinecarboxylate, Ethyl 2,4-dichloro-6-methylnicotinate
OR29728-5G
OR29728-25G
APOSOR29728-5GEA
79.9
EUR
APOSOR29728-5G
APOSOR29728-25G
Ethyl-2,4-dichloro-6-methylpyridine-3-carboxylate
Ethyl-2,4-dichloro-6-methylpyridine-3-carboxylate
Formel:
C₉H₉Cl₂NO₂ Molecular Weight: 234,08 g/mol Schmelzpunkt: 53…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173918 CAS-Nummer: 86129-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...