5-Methylisoxazole-3-carbothioamide

Lieferant: Apollo Scientific
Achtung

OR29729-1G OR29729-1MG OR29729-250MG OR29729-500MG OR29729-5G
APOSOR29729-1GEA 261 EUR
APOSOR29729-1G APOSOR29729-1MG APOSOR29729-250MG APOSOR29729-500MG APOSOR29729-5G
5-Methylisoxazole-3-carbothioamide
5-Methylisoxazole-3-carbothioamide
Formel: C₅H₆N₂OS
Molecular Weight: 142.18 g/mol
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173804
CAS-Nummer: 77358-26-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR