6-Methoxypyridazine-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR2975-1G
OR2975-250MG
OR2975-5G
APOSOR2975-1GEA
49.3
EUR
APOSOR2975-1G
APOSOR2975-250MG
APOSOR2975-5G
6-Methoxypyridazine-3-carboxylic acid
6-Methoxypyridazine-3-carboxylic acid
Formel:
C₆H₆N₂O₃ Schmelzpunkt: 160…163 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09800587 CAS-Nummer: 56434-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...