1-(2H-Pyrazol-3-yl)ethanone hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Acetyl-1H-pyrazole hydrochloride, 1-(1H-Pyrazol-3-yl)ethanone hydrochloride
OR29765-1G
OR29765-250MG
APOSOR29765-1GEA
89.9
EUR
APOSOR29765-1G
APOSOR29765-250MG
1-(2H-Pyrazol-3-yl)ethanone hydrochloride
1-(2H-Pyrazol-3-yl)ethanone hydrochloride
Formel:
C₅H₇ClN₂O Molecular Weight: 146.58 g/mol Schmelzpunkt: 150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204160 CAS-Nummer: 175277-40-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...