Methyl-6-chloropyridazine-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-chloropyridazine-3-carboxylate
OR2978-100MG
OR2978-1G
APOSOR2978-100MGEA
21.9
EUR
APOSOR2978-100MG
APOSOR2978-1G
Methyl-6-chloropyridazine-3-carboxylate
Methyl-6-chloropyridazine-3-carboxylate
Formel:
C₆H₅ClN₂O₂ Molecular Weight: 172,5691 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08694876 CAS-Nummer: 65202-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...