4-(4-Iodophenyl)morpholine

Lieferant: Apollo Scientific
Achtung

OR2980-1G OR2980-5G
APOSOR2980-1GEA 0 EUR
APOSOR2980-1G APOSOR2980-5G
4-(4-Iodophenyl)morpholine
4-(4-Iodophenyl)morpholine
Formel: C₁₀H₁₂INO
Schmelzpunkt: 161…163 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04974040
CAS-Nummer: 87350-77-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR