3-Chlorbenzhydrazid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Chlorobenzoic hydrazide
OR29831-100G
OR29831-25G
OR29831-5G
APOSOR29831-100GEA
458
EUR
APOSOR29831-100G
APOSOR29831-25G
APOSOR29831-5G
3-Chlorbenzhydrazid
3-Chlorbenzhydrazid
Formel:
C₇H₇ClN₂O Molecular Weight: 170.6 g/mol Schmelzpunkt: 156…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014757 CAS-Nummer: 1673-47-8 EINECS: 216-812-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...