2-Chlorisonicotinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure
OR29861-100G
OR29861-1G
OR29861-5G
APOSOR29861-100GEA
125
EUR
APOSOR29861-100G
APOSOR29861-1G
APOSOR29861-5G
2-Chlorisonicotinsäure
2-Chlorisonicotinsäure
Formel:
C₆H₄ClNO₂ Molecular Weight: 157.56 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191402 CAS-Nummer: 6313-54-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...