1-(3,5-Dichlor-4-pyridinyl)piperazin

Lieferant: Apollo Scientific
Gefahr

OR29877-1G OR29877-5G
APOSOR29877-1GEA 79.9 EUR
APOSOR29877-1G APOSOR29877-5G
1-(3,5-Dichlor-4-pyridinyl)piperazin
1-(3,5-Dichlor-4-pyridinyl)piperazin
Formel: C₉H₁₁Cl₂N₃
Molecular Weight: 232.11 g/mol
Schmelzpunkt: 75…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221400
CAS-Nummer: 175277-80-2
UN: 3259
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR