3-Chloro-5-methylpyridazine 97%

Lieferant: Apollo Scientific
Achtung

OR2988-1G OR2988-5G
APOSOR2988-1GEA 183 EUR
APOSOR2988-1G APOSOR2988-5G
3-Chloro-5-methylpyridazine 97%
3-Chloro-5-methylpyridazine
Formel: C₅H₅ClN₂
Lagertemperatur: Kühlschrank
MDL: MFCD09743645
CAS-Nummer: 89283-31-8
UN: 1593
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR