4-Brom-2,1,3-benzothiadiazol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromobenzo[c][1,2,5]thiadiazole

OR2989-1G
APOSOR2989-1GEA 79.9 EUR
APOSOR2989-1G
4-Brom-2,1,3-benzothiadiazol 95%
4-Bromo-2,1,3-benzothiadiazole
Formel: C₆H₃BrN₂S
Molecular Weight: 215.0704 g/mol
Schmelzpunkt: 80…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00614355
CAS-Nummer: 22034-13-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR