4-Methoxy-3-nitrobenzonitril 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Nitro-4-methoxybenzonitrile

OR29956-1G OR29956-25G OR29956-5G
APOSOR29956-1GEA 23.2 EUR
APOSOR29956-1G APOSOR29956-25G APOSOR29956-5G
4-Methoxy-3-nitrobenzonitril 95%
4-Methoxy-3-nitrobenzonitril
Formel: C₈H₆N₂O₃
Molecular Weight: 178,1448 g/mol
Schmelzpunkt: 148…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221439
CAS-Nummer: 33224-23-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR