6-Brom-1-chlorisochinolin

Lieferant: Apollo Scientific
Gefahr

OR300053-1G OR300053-5G OR300053-10G
APOSOR300053-1GEA 34.8 EUR
APOSOR300053-1G APOSOR300053-5G APOSOR300053-10G
6-Brom-1-chlorisochinolin
6-Bromo-1-chloroisoquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242,5000 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06738661
CAS-Nummer: 205055-63-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR