6-Bromo-3,4-dihydro-2H-isoquinolin-1-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-3,4-dihydroisoquinolin-1(2H)-one

OR300088-5G
APOSOR300088-5GEA 114 EUR
APOSOR300088-5G
6-Bromo-3,4-dihydro-2H-isoquinolin-1-one
6-Bromo-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₉H₈BrNO
Molecular Weight: 226,0700 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114858
CAS-Nummer: 147497-32-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR