6-Chloroisoquinoline
Lieferant: Apollo Scientific
|
Achtung
|
OR300089-1G
OR300089-5G
OR300089-10G
APOSOR300089-1GEA
21.9
EUR
APOSOR300089-1G
APOSOR300089-5G
APOSOR300089-10G
6-Chloroisoquinoline
6-Chloroisoquinoline
Formel:
C₉H₆ClN Molecular Weight: 163,6000 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09991646 CAS-Nummer: 62882-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...