3-Chloroisoquinolin-4-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-chloroisoquinoline
OR300097-500MG
OR300097-250MG
APOSOR300097-500MGEA
119
EUR
APOSOR300097-500MG
APOSOR300097-250MG
3-Chloroisoquinolin-4-amine
3-Chloroisoquinolin-4-amine
Formel:
C9H7ClN2 Molecular Weight: 178,6183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08235154 CAS-Nummer: 342899-38-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...