6-Bromo-8-chloroimidazo[1,2-a]pyrazine
Lieferant: Apollo Scientific
|
Achtung
|
OR300249-1G
OR300249-5G
OR300249-10G
APOSOR300249-1GEA
55.1
EUR
APOSOR300249-1G
APOSOR300249-5G
APOSOR300249-10G
6-Bromo-8-chloroimidazo[1,2-a]pyrazine
6-Bromo-8-chloroimidazo[1,2-a]pyrazine
Formel:
C₆H₃BrClN₃ Molecular Weight: 232,4700 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11846485 CAS-Nummer: 1208083-37-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...