Methyl-3-(phenylamino)propanoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 3-(phenylamino)propanoate
OR300505-1G
OR300505-5G
APOSOR300505-1GEA
132
EUR
APOSOR300505-1G
APOSOR300505-5G
Methyl-3-(phenylamino)propanoate
Methyl-3-(phenylamino)propanoate
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179,2200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00089334 CAS-Nummer: 21911-84-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...