tert-Butyl-2-(benzylamino)acetate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Benzylglycine tert-butyl ester
OR300515-1G
OR300515-5G
APOSOR300515-1GEA
0
EUR
APOSOR300515-1G
APOSOR300515-5G
tert-Butyl-2-(benzylamino)acetate
tert-Butyl-2-(benzylamino)acetate
Formel:
C₁₃H₁₉NO₂ Molecular Weight: 221,3 g/mol Siedepunkt: 125…126 °C/0,5mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD11848800 CAS-Nummer: 7662-76-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...