2-Chloro-4-(3-nitrophenyl)[1,3,5]triazine

Lieferant: Apollo Scientific

OR300585-1G OR300585-250MG OR300585-500MG OR300585-2.5G
APOSOR300585-1GEA 0 EUR
APOSOR300585-1G APOSOR300585-250MG APOSOR300585-500MG APOSOR300585-2.5G
2-Chloro-4-(3-nitrophenyl)[1,3,5]triazine
2-Chloro-4-(3-nitrophenyl)[1,3,5]triazine
Formel: C₉H₅ClN₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11052416
CAS-Nummer: 919085-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR