6-(Methoxycarbonyl)nicotinic acid

Lieferant: Apollo Scientific
Achtung

OR300619-1G OR300619-5G
APOSOR300619-1GEA 79.9 EUR
APOSOR300619-1G APOSOR300619-5G
6-(Methoxycarbonyl)nicotinic acid
6-(Methoxycarbonyl)nicotinic acid
Formel: C₈H₇NO₄
Schmelzpunkt: 220…222 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09702182
CAS-Nummer: 17874-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR