2-Chloro-4-phenylnicotinonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-3-cyano-4-phenylpyridine

OR300654-5G
APOSOR300654-5GEA 0 EUR
APOSOR300654-5G
2-Chloro-4-phenylnicotinonitrile 97%
2-Chloro-4-phenylnicotinonitrile
Formel: C₁₂H₇ClN₂
Molecular Weight: 214,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01935958
CAS-Nummer: 163563-64-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR