5-(4-Bromophenyl)cyclohexane-1,3-dione
Lieferant: Apollo Scientific
OR300683-1G
OR300683-250MG
OR300683-500MG
OR300683-5G
APOSOR300683-1GEA
288
EUR
APOSOR300683-1G
APOSOR300683-250MG
APOSOR300683-500MG
APOSOR300683-5G
5-(4-Bromophenyl)cyclohexane-1,3-dione
5-(4-Bromophenyl)cyclohexane-1,3-dione
Formel:
C₁₂H₁₁BrO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03844400 CAS-Nummer: 239132-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...