6-Hydroxymethylnicotinic acid ethyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 6-(hydroxymethyl)pyridine-3-carboxylate

OR300696-1G OR300696-500MG OR300696-5G
APOSOR300696-1GEA 0 EUR
APOSOR300696-1G APOSOR300696-500MG APOSOR300696-5G
6-Hydroxymethylnicotinic acid ethyl ester
6-Hydroxymethylnicotinic acid ethyl ester
Formel: C₉H₁₁NO₃
Molecular Weight: 181,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12131130
CAS-Nummer: 35005-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR