4-Nitro-1-isoindolinone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitroisoindolin-1-one

OR300701-1G OR300701-250MG OR300701-5G
APOSOR300701-1GEA 109 EUR
APOSOR300701-1G APOSOR300701-250MG APOSOR300701-5G
4-Nitro-1-isoindolinone 98%
4-Nitro-1-isoindolinone
Formel: C₈H₆N₂O₃
Molecular Weight: 178,15 g/mol
Schmelzpunkt: 225…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09701290
CAS-Nummer: 366452-97-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR