2,3,5-Trichlorbenzaldehyd 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR300750-25G
OR300750-5G
APOSOR300750-25GEA
337
EUR
APOSOR300750-25G
APOSOR300750-5G
2,3,5-Trichlorbenzaldehyd 98%
2,3,5-Trichlorobenzaldehyde
Formel:
C₇H₃Cl₃O Molecular Weight: 209,46 g/mol Schmelzpunkt: 75…76 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010860 CAS-Nummer: 56961-75-2 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...