2-Phenylaminomalonic acid diethyl ester 95+%
Lieferant: Apollo Scientific
Synonyme: Diethyl 2-(phenylamino)malonate
OR300758-1G OR300758-5G
APOSOR300758-1GEA 0 EUR
APOSOR300758-1G APOSOR300758-5G
2-Phenylaminomalonic acid diethyl ester 95+%
2-Phenylaminomalonic acid diethyl ester
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00122281
CAS-Nummer: 6414-58-0