Methyl-2-(brommethyl)-3-nitrobenzoat 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl 2-(bromomethyl)-3-nitrobenzoate
OR300783-25G
OR300783-100G
APOSOR300783-25GEA
59.6
EUR
APOSOR300783-25G
APOSOR300783-100G
Methyl-2-(brommethyl)-3-nitrobenzoat 95%
Methyl-2-(brommethyl)-3-nitrobenzoat
Formel:
C₉H₈BrNO₄ Molecular Weight: 274.07 g/mol Schmelzpunkt: 75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114315 CAS-Nummer: 98475-07-1 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...