Methylaminophenylacetonitrile hydrochloride

Lieferant: Apollo Scientific

OR300787-10G OR300787-1G OR300787-5G
APOSOR300787-10GEA 0 EUR
APOSOR300787-10G APOSOR300787-1G APOSOR300787-5G
Methylaminophenylacetonitrile hydrochloride
Methylaminophenylacetonitrile hydrochloride
Formel: C₉H₁₁ClN₂
Molecular Weight: 182,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11052442
CAS-Nummer: 5537-52-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR