(2,4-Dioxothiazolidin-5-yl)acetic acid
Lieferant: Apollo Scientific
Synonyme:
2-(2,4-Dioxothiazolidin-5-yl)acetic acid
OR300906-1G
OR300906-250MG
OR300906-500MG
APOSOR300906-1GEA
221
EUR
APOSOR300906-1G
APOSOR300906-250MG
APOSOR300906-500MG
(2,4-Dioxothiazolidin-5-yl)acetic acid
(2,4-Dioxothiazolidin-5-yl)acetic acid
Formel:
C₅H₅NO₄S Molecular Weight: 175.16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01165868 CAS-Nummer: 875-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...