2-Chlorobenzo[d]thiazol-6-amine 97%

Lieferant: Apollo Scientific

Synonyme: 6-Amino-2-chlorobenzothiazole

OR300915-1G OR300915-5G
APOSOR300915-1GEA 0 EUR
APOSOR300915-1G APOSOR300915-5G
2-Chlorobenzo[d]thiazol-6-amine 97%
2-Chlorobenzo[d]thiazol-6-amine
Formel: C₇H₅ClN₂S
Molecular Weight: 184.646 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08276901
CAS-Nummer: 2406-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR