3-Brom-o-kresol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Bromo-2-methylphenol
OR301014-5G
OR301014-25G
APOSOR301014-5GEA
72.5
EUR
APOSOR301014-5G
APOSOR301014-25G
3-Brom-o-kresol
3-Brom-o-kresol
Formel:
C₇H₇BrO Molecular Weight: 187,0339 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100990 CAS-Nummer: 7766-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...