1-Brom-5-chlor-2-nitrobenzol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-chloro-1-nitrobenzene
OR301067-25G
OR301067-5G
APOSOR301067-25GEA
48
EUR
APOSOR301067-25G
APOSOR301067-5G
1-Brom-5-chlor-2-nitrobenzol 97%
1-Brom-5-chlor-2-nitrobenzol
Formel:
C₆H₃BrClNO₂ Molecular Weight: 236,4500 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08437614 CAS-Nummer: 63860-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...