3,5-Dichlorpyridazin
Lieferant: Apollo Scientific
|
Achtung
|
OR301078-1G
OR301078-25G
OR301078-5G
APOSOR301078-1GEA
23.2
EUR
APOSOR301078-1G
APOSOR301078-25G
APOSOR301078-5G
3,5-Dichlorpyridazin
3,5-Dichloropyridazine
Formel:
C₄H₂Cl₂N₂ Molecular Weight: 148.9781 g/mol Schmelzpunkt: 58…64 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10698048 CAS-Nummer: 1837-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...