2-Amino-1,3,5-triazine-4,6-dithiol 90%
Lieferant: Apollo Scientific
|
Achtung
|
OR30109-5G
OR30109-1G
APOSOR30109-5GEA
125
EUR
APOSOR30109-5G
APOSOR30109-1G
2-Amino-1,3,5-triazine-4,6-dithiol 90%
2-Amino-1,3,5-triazine-4,6-dithiol
Formel:
C₃H₄N₄S₂ Molecular Weight: 160,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00047346 CAS-Nummer: 2770-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...