6,7-Dihydro-5H-quinoline-8-one

Lieferant: Apollo Scientific
Achtung

OR301103-1G OR301103-5G
APOSOR301103-1GEA 21.9 EUR
APOSOR301103-1G APOSOR301103-5G
6,7-Dihydro-5H-quinoline-8-one
6,7-Dihydro-5H-quinoline-8-one
Formel: C₉H₉NO
Molecular Weight: 147,1700 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03839916
CAS-Nummer: 56826-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR