6-Aminomethylquinoline 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-(Aminomethyl)quinoline

OR301111-1G OR301111-250MG OR301111-5G
APOSOR301111-1GEA 143 EUR
APOSOR301111-1G APOSOR301111-250MG APOSOR301111-5G
6-Aminomethylquinoline 95%
6-Aminomethylquinoline
Formel: C₁₀H₁₀N₂
Molecular Weight: 158,204 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD02853688
CAS-Nummer: 99071-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR