7-Chinolinol 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Quinolin-7-ol, 7-Hydroxyquinoline
OR301114-5G
OR301114-1G
OR301114-10G
APOSOR301114-5GEA
63.8
EUR
APOSOR301114-5G
APOSOR301114-1G
APOSOR301114-10G
7-Chinolinol 98%
7-Chinolinol
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 239…244 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016730 CAS-Nummer: 580-20-1 EINECS: 209-457-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...