1-Methylbenzoimidazol-5-amine

Lieferant: Apollo Scientific
Achtung

OR301135-1G OR301135-250MG
APOSOR301135-1GEA 0 EUR
APOSOR301135-1G APOSOR301135-250MG
1-Methylbenzoimidazol-5-amine
1-Methylbenzoimidazol-5-amine
Formel: C₈H₉N₃
Schmelzpunkt: 156…159 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03164349
CAS-Nummer: 10394-38-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR