6-Bromo-1H-indazole-4-carboxylic acid

Lieferant: Apollo Scientific
Gefahr

OR301140-10G OR301140-1G OR301140-5G
APOSOR301140-10GEA 145 EUR
APOSOR301140-10G APOSOR301140-1G APOSOR301140-5G
6-Bromo-1H-indazole-4-carboxylic acid
6-Bromo-1H-indazole-4-carboxylic acid
Formel: C₈H₅BrN₂O₂
Molecular Weight: 241,0400 g/mol
Schmelzpunkt: 293…298 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07368215
CAS-Nummer: 885523-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR